Få mere at vide
Tocris Bioscience™ NSC 668036
GREENER_CHOICE
Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein
Brand:  Tocris Bioscience™ 5813/10
Encompass
Produktkode 15767126
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
NSC 668036 is a Wnt signaling inhibitor. Binds the PDZ domain of the Dishevelled (Dvl) protein. Inhibits proliferation, migration and TGF-β-induced differentiation of fibroblasts in vitro. Reduces severity of pulmonary fibrosis in a mouse model.
Tekniske data
Tekniske data
N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-alanine-(1S)-1-carboxy-2-methylpropyl ester
10 mg
Solid
Bioactive Small Molecule
144678-63-7
Soluble to 100mM in DMSO and to 100mM in ethanol
White
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only.