Få mere at vide
Tocris Bioscience™ NSC 636819
Beskrivelse
NSC 636819 is a KDM4A/KDM4B inhibitor; inhibits histone demethylase activity on H3K9me3 and induces apoptosis in LNCaP prostate cancer cells in vitro. Also negatively regulates androgen-responsive genes.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,5-Bis[(1E)-2-(3,4-dichlorophenyl)ethenyl]-2,4-dinitrobenzene |
| CAS | 1618672-71-1 |
| Mængde | 50 mg |
| Mål | Histone Demethylase Inhibitors |
| Molekylær formel | C22H12Cl4N2O4 |
| Formel vægt | 510.15 |
| Renhed | >98% |