Få mere at vide
Tocris Bioscience™ NSC 617145
GREENER_CHOICE
Werner syndrome helicase (WRN) inhibitor
Brand:  Tocris Bioscience™ 5340/50
Encompass
Produktkode 15706736
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
NSC 617145 is a werner syndrome helicase (WRN) helicase inhibitor (IC50 = 250 nM); acts synergistically with Mitomycin C (Cat. No. 3258) to induce double-strand breaks and chromosomal abnormalities in vitro.
Tekniske data
Tekniske data
NSC 617145
1,1'-(2,2-Dimethyl-1,3-propanediyl)bis[3,4-dichloro-1H-pyrrole-2,5-dione
Soluble to 20mM in DMSO
Bioactive Small Molecule
203115-63-3
50 mg
Solid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
Research Use Only. Not For Human or Veterinary Use