Få mere at vide
Tocris Bioscience™ NS 19504
GREENER_CHOICE
KCa1.1 (BK) channel activator
Brand:  Tocris Bioscience™ 5276/50
Encompass
Produktkode 15776646
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
NS 19504 is a selective activator of large-conductance Ca2+-activated potassium channels (BKCa, KCa1.1) (EC50 = 11 μM). Activates BK channels in native smooth muscle cells from guinea pig bladder and reduces spontaneous phasic contractions in bladder strips ex vivo. Displays no effect on contractions induced by a high K+ concentration or at 65 other receptors tested, including L- and N- type Ca2+ channels, Nav, KATP and hERG channels.
Tekniske data
Tekniske data
5-[(4-Bromophenyl)methyl]-2-thiazolamine
327062-46-4
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner