missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRF3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05800AF532
This item is not returnable.
View return policy
Description
NRF3 Polyclonal specifically detects NRF3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
NRF3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
NF-E2-related factor 3, NFE2-related factor 3, Nrf3, NRF3nuclear factor erythroid 2-related factor 3, nuclear factor (erythroid-derived 2)-like 3, Nuclear factor, erythroid derived 2, like 3, nuclear factor-erythroid 2 p45-related factor 3, nuclear factor-erythroid 2-related factor 3 | |
Produced in rabbits immunized with E. coli-derived Human NRF3 fragment. | |
0.1 mL | |
Stem Cell Markers | |
9603 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction