missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ NPM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116760
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Positive Samples: HeLa, Raji Immunogen sequence: APVTMDSFFF GCELSGHTRS FTFKVEEEDD AEHVLALTML CLTEGAKDEC NVVEVVARNH DHQEIAVPVA NLKLSCQPML SLDDFQLQPP VTFRLKSGSG PVRITGRHQI V.
The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus.
Tekniske data
| NPM3 | |
| Polyclonal | |
| Unconjugated | |
| NPM3 | |
| Npm3; Nub1; nucleophosmin/nucleoplasmin 3; nucleophosmin/nucleoplasmin family, member 3; nucleophosmin/nucleoplasmin, 3; nucleoplasmin 3; nucleoplasmin-3; PORMIN; RGD1565542; TMEM123 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10360 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75607 | |
| NPM3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human NPM3 (NP_0089241). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion