Få mere at vide
NPG Glycol-d6, TRC
Artikelnummer. 30414426
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30414426 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414426 Leverandør Toronto Research Chemicals Leverandørnr. TRCN897006500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2-Dimethyl-1,3-propanediol D6 (2,2-Dimethyl D6)
CAS 14671-00-2
Molekylær formel C5 D6 H6 O2
Mængde 500 mg
Synonym 2,2-Dimethyl-1,3-propanediol D6 (2,2-Dimethyl D6), 2,2-Di(methyl-d3)-1,3-propanediol
InChI formel InChI=1S/C5H12O2/c1-5(2,3-6)4-7/h6-7H,3-4H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])C(CO)(CO)C([2H])([2H])[2H]
IUPAC navn 2,2-bis(trideuteriomethyl)propane-1,3-diol
Molekylvægt (g/mol) 110.185
Formel vægt 110.121