Få mere at vide
NPEC-caged-noradrenalin, TRC
Artikelnummer. 30411565
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30411565 1 mg -
30414093 10 mg -
30411999 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30411565 Leverandør Toronto Research Chemicals Leverandørnr. TRCN8990201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1257323-83-3
Molekylær formel C17H18N2O7
Molekylvægt (g/mol) 362.33
SMIL O=C(OC(C1=CC=CC=C1[N+]([O-])=O)C)NC[C@@H](C2=CC=C(O)C(O)=C2)O
Tekniske data
Kemisk navn eller materiale NPEC-caged-noradrenalin
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C17H18N2O7/c1-10(12-4-2-3-5-13(12)19(24)25)26-17(23)18-9-16(22)11-6-7-14(20)15(21)8-11/h2-8,10,16,20-22H,9H2,1H3,(H,18,23)/t10?,16-/m0/s1
Formel vægt 362.1114
Procent renhed >95%