Få mere at vide
Noxiptillin Hydrochloride, LoGiCal
Artikelnummer. 30461334
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461334 10 mg
1 options
Artikelnummer. 30461334 Leverandør LoGiCal Leverandørnr. LGCFOR3163.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Noxiptillin Hydrochloride
CAS 4985-15-3
Analyt- eller komponentnavne Noxiptillin Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C19 H22 N2 O . Cl H
Certificeringer/compliance ISO 9001
Synonym 5H-Dibenzo[a,d]cyclohepten-5-one, 10,11-dihydro-, O-[2-(dimethylamino)ethyl]oxime, hydrochloride (1:1), 5H-Dibenzo[a,d]cyclohepten-5-one, 10,11-dihydro-, O-[2-(dimethylamino)ethyl]oxime, monohydrochloride (9CI), Agedal, BAY 1521, CD 37B, Nogedal, Noxiptilin hydrochloride, Noxiptiline hydrochloride, O-(2-Dimethylaminoethyl)-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-one oxime hydrochloride
InChI formel InChI=1S/C19H22N2O.ClH/c1-21(2)13-14-22-20-19-17-9-5-3-7-15(17)11-12-16-8-4-6-10-18(16)19;/h3-10H,11-14H2,1-2H3;1H
Løsningstype Neat
SMIL Cl.CN(C)CCON=C1c2ccccc2CCc3ccccc13
IUPAC navn 2-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylideneamino)oxy-N,N-dimethylethanamine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 330.85
Mængde 10 mg
Formel vægt 330.1499 g/mol