missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human AKT (Total) ELISA Kit

Artikelnummer. 10364403
Klik for at se tilgængelige muligheder
Mængde:
96 prøver
Pakningsstørrelse:
96 tests
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 10364403

Brand: Invitrogen™ KHO0101

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

ELISA

Kit is designed to detect and quantify the level of AKT (Total) in fresh or frozen human cell lysates. Cross-reactivity in mouse and rat cell lysates has been observed. The assay recognizes both natural and recomb. AKT (Total), independent of its phosphorylation state. Principle of the method A monoclonal capture antibody specific for AKT (Total) has been coated onto the wells of 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a HRP labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

AKT også kendt som proteinkinase B (PKB) eller RAS-alpha, er en allestedsnærværende serin/threoninkinase, der spiller en vigtig rolle i forskellige biologiske responser såsom regulering af metabolisme, celleoverlevelse og vækst ved at fosforylere flere proteiner. Denne proteinkinase aktiveres af insulin, PI3K, IGF1 og forskellige andre vækst- og overlevelsesfaktorer. Akt fremmer celleoverlevelse ved at hæmme apoptose gennem phosphorylering og inaktivering af flere mål, herunder gaffelhoved-transskriptionsfaktorer og caspase-9. AKT-vejen er et vigtigt mål for opdagelse af kræftlægemidler.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P31749,P31750,P31751,P47196,P47197,Q60823,Q63484,Q9WUA6,Q9Y243
Analyseområde 0.3 to 20 ng/mL
Assay følsomhed <0.1 ng/mL
Koncentration 0.3 to 20 ng/mL
Konjugeret HRP
Produkttype ELISA
Prøvetype Cellelysater
Substrattype Serine/Threonine Kinases
Målarter Mus, Rotte
Til brug med (udstyr) Kolorimetrisk mikropladelæser
Gen-id (Entrez) 10000, 11651, 11652, 207, 208, 23797, 24185, 25233, 29414
Gen symbol AKT,CWS6,PKB,PKB-ALPHA,PRKBA,RAC,RAC-ALPHA,HIHGHH,PKBB,PKBBETA,PRKBB,RAC-BETA,MPPH,MPPH2,PKB-GAMMA,PKBG,PRKBG,RAC-Pk.-gamma,RAC-gamma,STK-2
Inkubationstid 4 hr.
Interassay CV 9.3%
Intraassay CV 7.7
Sættets indhold Forbelagt 96-brønds plade, Standard, Standardfortyndingsbuffer, Detektionsantistof, anti-rabbit-HRP, HRP fortynder, Vaskebuffer, kromogen, Stop Løsning, Klæbende pladedæksler
Etiket eller farvestof HRP
Markør Gene Akt
Mængde 96 prøver
Regulatorisk status RUO
Prøvevolumen Cell Lysate 10 μL
Opbevaringskrav 2 °C til 8 °C
Mål AKT Pan
Test tid 1 t 20 min
Samlet analysetid 4 hrs
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.