Learn More
Invitrogen™ IP-10 Monkey Singleplex Magnetic Bead Kit
Designed for the measurement of IP-10 (Interferon-gamma-inducible, 10 kD protein) in Rhesus monkey and Cynomolgus monkey serum, plasma, and tissue culture supernatant
Brand: Invitrogen™ LPC1171M
Additional Details : Weight : 0.01000kg
Includes: Capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet.
Requires : Buffer reagents needed to complete the reaction are sold separately under Catalog number LHB0001.
Description
Assays using the IP-10 Monkey Singleplex Magnetic Bead Kit may be run alone or in combination with the Monkey Cytokine Magnetic 28-Plex Panel. Kit employs magnetic beads, facilitating automation, decreasing hands-on time, and increasing throughput and precision. The panel is suitable for use with the Luminex™ 200™ FLEXMAP 3D™ MAGPIX systems.
- Superior performance—accurate, reproducible, and sensitive quantitation of selected proteins
- High quality—fully qualified antibodies permit excellent specificity and sensitivity
- Fast and easy protocols—perform your assays and analyze your data typically in less than one day
The IP-10 Monkey Singleplex Magnetic Bead Kit is based on xMAP technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex assays. Magnetic microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. At the completion of the sandwich immunoassay, beads can be read using the Luminex 200™ FLEXMAP 3D™ detection systems, instruments based on flow cytometry, two lasers, real-time digital signal processing, and xPONENT™ software to distinguish bead color (analyte) and assay signal strength (PE) fluorescence intensity.
MAGPIX™ System—Brings the Power of Multiplex to Every Lab system may also be used for sample analysis. This instrument utilizes xMAP technology and is built on light-emitting diodes (LEDs) for excitation, a CCD camera for bead and analyte detection and high-speed digital signal processors. The MAGPIX™ system is more compact and robust, providing a very cost effective magnetic bead-based multiplexing tool. Simple, out of the box set-up allows researchers to rapidly perform singleplex or multiplex assays of up to 50 targets in single biological samples. The MAGPIX™ system is easy to use—ideal for both new and experienced users alike.
Magnetic Beads Let You Do More with Your Time
Magnetic bead-based assays use Luminex™ MagPlex magnetic microsphere. All other assay components are the same as the equivalent Luminex™ polystyrene bead-based assays providing identical quality and consistency performance. MagPlex technology takes advantage of magnetic properties to simplify assay wash steps and maximize uniformity of results. Magnetic bead-based assays also enable automation, decrease hands-on time, and increase throughput and precision. Easy to use protocols allow you to get your results in as little as 3.5 hours. In addition, these assays are compatible with both vacuum and magnetic wash stations and work well with the MAGPIX system as well as other Luminex xMAP platforms.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
1 Panel | |
IP-10 | |
Capture antibody coated beads, Detector antibody, Lot specific technical data sheet, Standard |
Fluorescent Detection | |
Serum |
For Research Use Only. Not for use in diagnostic procedures.