Få mere at vide
Emma Safety Footwear Novara sikkerhedssko
Størrelse:
39
40
41
42
43
44
45
46
47
48
Pakningsstørrelse:
1 par
Beskrivelse
- S3-Safety shoe with sole plate for wet work
- Full grain leather / Nubuck construction
- Chemical resistant / Liquid repellant
- Nitrile Rubber Heat Resistant Outsole
- ESD certified
- Chrome-free lining
- Sanitized
Tekniske data
Tekniske data
| Certificeringer/compliance | EN ISO 20345:2011, EN 12568:2010 |
| Lukketype | Laces |
| Farve | Brown |
| Design | Novara |
| Liner materiale | Hydro-Tec™ Sanitized Silver & Wellness leather |
| Materiale | Leather, Nubuck |
| Størrelse | 40 |
| Tå materiale | Steel |