Få mere at vide
Novaluron, TRC
Artikelnummer. 30427078
Skift visning
Mængde:
50 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427078 50 mg
30410684 500 mg
30415037 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427078 Leverandør Toronto Research Chemicals Leverandørnr. TRCN88840050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 116714-46-6
Molekylær formel C17 H9 Cl F8 N2 O4
Molekylvægt (g/mol) 492.7
Synonym Rimon, 1-[3-chloro-4-(1,1,2-trifluoroethoxy)phenyl]-3-(2,6-difluorobenzoyl)urea
IUPAC navn N-[[3-chloro-4-[1,1,2-trifluoro-2-(trifluoromethoxy)ethoxy]phenyl]carbamoyl]-2,6-difluorobenzamide
SMIL FC(OC(F)(F)F)C(F)(F)Oc1ccc(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl
Tekniske data
Kemisk navn eller materiale Novaluron
Mængde 50 mg
InChI formel InChI=1S/C17H9ClF8N2O4/c18-8-6-7(4-5-11(8)31-16(22,23)14(21)32-17(24,25)26)27-15(30)28-13(29)12-9(19)2-1-3-10(12)20/h1-6,14H,(H2,27,28,29,30)
Formel vægt 492.0123