missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Novagen™ rLysozyme™ Solution

For lysis of Gram-negative bacteria

POA


Description

Description

  • Contains highly purified and stabilized recombinant lysozyme
  • Catalyzes hydrolysis of N-acetylmuramide linkages in bacterial cell walls
  • Specific activity of rLysozyme (1700KU/mg) for E. coli lysis is 250 times greater than that of chicken egg white lysozyme
  • Optimally active at physiological pH
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only