Få mere at vide
Cytiva NovaBlot Kit
Shop alle Cytiva produkter
Type:
Elektroblottingsæt
Pakningsstørrelse:
1 sæt
Beskrivelse
Indeholder
- 1 x Electrode cathode
- 1 x Electrode anode
- 500 x Electrode paper (200x250mm)
- 50 x Cellophane sheet (210x320mm)
- For use with Multiphor II
- Build sandwiches to simultaneously transfer from up to 6 gels at a time
- Transfer usually takes less than an hour and does not require a cooling period at the end of the transfer
- Electrodes included in the kit are made of graphite to provide an even distribution of current across the entire surface
- Maximum gel size: 200 x 250mm
- Electrophoretic transfer
- General protein staining
- Immunoblotting
- Glycoprotein detection
Tekniske data
Tekniske data
| Til brug med (udstyr) | Multiphor™ II Electrophoresis System |
| Type | Elektroblottingsæt |
| Inkluderer | Cathode and anode electrode, 500 electrode Paper (200 x 250mm), 50 cellophane sheet (210 x 320mm), Graphite electrode |