Få mere at vide
Nortriptyline-d3 Hydrochloride, TRC
Artikelnummer. 30412263
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30412263 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412263 Leverandør Toronto Research Chemicals Leverandørnr. TRCN8370021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nortriptyline-D3 Hydrochloride
CAS 203784-52-5
Molekylær formel C19 2H3 H18 N . Cl H
Mængde 1 mg
Synonym Nortriptyline-D3 Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-1-propanamine hydrochloride, 1-Propanamine','3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-','hydrochloride (1:1), 1-Propanamine','3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-','hydrochloride (9CI), 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-1-propanamine hydrochloride (1:1)
InChI formel InChI=1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20H,6,12-14H2,1H3;1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])NCCC=C1c2ccccc2CCc3ccccc13
IUPAC navn 3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-N-(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 302.86
Formel vægt 302.1629