Få mere at vide
Noroxymorphone Hydrochloride, Mikromol™
Artikelnummer. 17991326
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991326 100 mg
17983806 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17991326 Leverandør Mikromol Leverandørnr. MM0673.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Noroxymorphone Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Noroxymorphone Hydrochloride
CAS 52446-24-9
Analyt- eller komponentnavne Noroxymorphone Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H17 N O4 . Cl H
Certificeringer/compliance ISO 17025
Synonym Naloxone Hydrochloride Dihydrate Imp. A (EP), Naloxone Imp. A (EP), Noroxymorphone Hydrochloride, Naloxone Hydrochloride Dihydrate Impurity A as Hydrochloride, Naltrexone Hydrochloride Impurity B as Hydrochloride, Naloxone Impurity A as Hydrochloride, Naltrexone Impurity B as Hydrochloride
InChI formel InChI=1S/C16H17NO4.ClH/c18-9-2-1-8-7-11-16(20)4-3-10(19)14-15(16,5-6-17-11)12(8)13(9)21-14;/h1-2,11,14,17-18,20H,3-7H2;1H/t11-,14+,15+,16-;/m1./s1
SMIL Cl.Oc1ccc2C[C@H]3NCC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O
IUPAC navn (4R,4aS,7aR,12bS)-4a,9-dihydroxy-1,2,3,4,5,6,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
Molekylvægt (g/mol) 323.77
Mængde 100 mg
Formel vægt 323.0924 g/mol