Få mere at vide
(-)-Normacromerine-d3, TRC
Artikelnummer. 30426644
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30426644 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426644 Leverandør Toronto Research Chemicals Leverandørnr. TRCN71700250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (-)-Normacromerine-d3
CAS 1329805-62-0
Molekylær formel C11H14D3NO3
Mængde 50 mg
Synonym (alphaR)-3,4-Dimethoxy-alpha-[(methylamino)methyl]benzenemethanol-d3, Normacromerine-d3
InChI formel InChI=1S/C11H17NO3/c1-12-7-9(13)8-4-5-10(14-2)11(6-8)15-3/h4-6,9,12-13H,7H2,1-3H3/t9-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])NC[C@H](O)c1ccc(OC)c(OC)c1
IUPAC navn (1R)-1-(3,4-dimethoxyphenyl)-2-(trideuteriomethylamino)ethanol
Molekylvægt (g/mol) 214.28
Formel vægt 214.1397