Få mere at vide
Norflurazon-13C,d3, TRC
Artikelnummer. 30418509
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30418509 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418509 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68450210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Norflurazon-13C,d3
CAS 1346603-47-1
Molekylær formel C1113CH6D3ClF3N3O
Mængde 10 mg
Synonym 4-Chloro-5-(methylamino-13C,d3)-2-[3-(trifluoromethyl)phenyl]-3(2H)-pyridazinone, 1-(3-Trifluoromethylphenyl)-4-(methylamino-13C,d3)-5-chloro-6-pyridazone, 4-Chloro-5-(methylamino-13C,d3)-2-(α,α,α-trifluoro-m-tolyl)-3(2H)-pyridazinone, H 9789-13C,d3, Predict-13C,d3, SAN 978938-13C,d3, Sandoz 9789-13C,d3, Solicam-13C,d3, Zorial-13C,d3
InChI formel InChI=1S/C12H9ClF3N3O/c1-17-9-6-18-19(11(20)10(9)13)8-4-2-3-7(5-8)12(14,15)16/h2-6,17H,1H3/i1+1D3
SMIL ClC1=C(N[13C]([2H])([2H])[2H])C=NN(C2=CC=CC(C(F)(F)F)=C2)C1=O
Molekylvægt (g/mol) 307.68
Formel vægt 307.0608