Få mere at vide
Norfluoxetine-d5 Hydrochloride, TRC
Artikelnummer. 30420187
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30420187 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420187 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68110210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Norfluoxetine-d5 Hydrochloride
CAS 1185132-92-6
Renhedsgrad noter HPLC
Molekylær formel C16 D5 H11 F3 N O . Cl H
Mængde 10 mg
Synonym 1,1,2,2,3-Pentadeuterio-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine hydrochloride, Norfluoxetine-d5 hydrochloride, Norfluoxetine-D5 (propyl-D5) Hydrochloride, γ-[4-(Trifluoromethyl)phenoxy]benzenepropan-α,α,β,β,γ-d5-amine hydrochloride (1:1), 3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1,1,2,2,3-d5-1-amine hydrochloride (1:1), 3-(4-Trifluoromethylphenoxy)-3-phenylpropyl-d5-amine hydrochloride, 1-(3-Amino-1-phenylpropoxy-1,2,2,3,3-d5)-4-(trifluoromethyl)benzene hydrochloride, (±)-Norfluoxetine-d5 HCl (propyl-1,1,2,2,3-d5)
InChI formel InChI=1S/C16H16F3NO.ClH/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;/h1-9,15H,10-11,20H2;1H/i10D2,11D2,15D;
SMIL Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])(Oc1ccc(cc1)C(F)(F)F)c2ccccc2
IUPAC navn 1,1,2,2,3-pentadeuterio-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Molekylvægt (g/mol) 336.7913
Formel vægt 336.1265
Procent renhed >95