missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norfluoxetine-d5 Hydrochloride, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Norfluoxetine-d5 Hydrochloride |
| CAS | 1185132-92-6 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C16 D5 H11 F3 N O . Cl H |
| Mængde | 1 mg |
| Synonym | 1,1,2,2,3-Pentadeuterio-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine hydrochloride, Norfluoxetine-d5 hydrochloride, Norfluoxetine-D5 (propyl-D5) Hydrochloride, γ-[4-(Trifluoromethyl)phenoxy]benzenepropan-α,α,β,β,γ-d5-amine hydrochloride (1:1), 3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1,1,2,2,3-d5-1-amine hydrochloride (1:1), 3-(4-Trifluoromethylphenoxy)-3-phenylpropyl-d5-amine hydrochloride, 1-(3-Amino-1-phenylpropoxy-1,2,2,3,3-d5)-4-(trifluoromethyl)benzene hydrochloride, (±)-Norfluoxetine-d5 HCl (propyl-1,1,2,2,3-d5) |
| InChI formel | InChI=1S/C16H16F3NO.ClH/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;/h1-9,15H,10-11,20H2;1H/i10D2,11D2,15D; |
| SMIL | Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])(Oc1ccc(cc1)C(F)(F)F)c2ccccc2 |
| IUPAC navn | 1,1,2,2,3-pentadeuterio-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride |
| Molekylvægt (g/mol) | 336.7913 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?