Få mere at vide
(±)-Norfluoxetine-d5 HCl (propyl-1,1,2,2,3-d5), CDN
Artikelnummer. 30322684
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322684 0.01 g
30323110 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322684 Leverandør CDN Leverandørnr. CDND60230.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Norfluoxetine-d5 Hydrochloride
CAS 1185132-92-6
Molekylær formel C16 D5 H11 F3 N O . Cl H
Mængde 0.01 g
Synonym 1,1,2,2,3-Pentadeuterio-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine hydrochloride, Norfluoxetine-d5 hydrochloride, Norfluoxetine-D5 (propyl-D5) Hydrochloride, γ-[4-(Trifluoromethyl)phenoxy]benzenepropan-α,α,β,β,γ-d5-amine hydrochloride (1:1), 3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1,1,2,2,3-d5-1-amine hydrochloride (1:1), (±)-Norfluoxetine-d5 HCl (propyl-1,1,2,2,3-d5), (±)-Norfluoxetine-d5 HCl (propyl-1,1,2,2,3-d5), (±)-Norfluoxetine-d5 HCl (propyl-1,1,2,2,3-d5)
InChI formel InChI=1 S/C16H16F3NO.ClH/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;/h1-9,15 H,10-11,20H2;1 H/i10D2,11D2,15 D;
SMIL Cl.[2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])(Oc1ccc(cc1)C(F)(F)F)c2ccccc2
IUPAC navn 1,1,2,2,3-pentadeuterio-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Molekylvægt (g/mol) 336.7913
Formel vægt 336.1265 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity