Få mere at vide
Norfloxacin-d8, TRC
Artikelnummer. 30424535
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30424535 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424535 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68100225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Norfloxacin D8 (piperazine D8)
CAS 1216601-32-9
Molekylær formel C16 2H8 H10 F N3 O3
Mængde 25 mg
Synonym Norfloxacin-d8 (piperazine-d8), 1-Ethyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxo-quinoline-3-carboxylic acid, Norfloxacin D8 (piperazine D8)
InChI formel InChI=1S/C16H18FN3O3/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20/h7-9,18H,2-6H2,1H3,(H,22,23)/i3D2,4D2,5D2,6D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(c2cc3N(CC)C=C(C(=O)O)C(=O)c3cc2F)C1([2H])[2H]
IUPAC navn 1-ethyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
Molekylvægt (g/mol) 327.38
Formel vægt 327.1834