Få mere at vide
Norfenfluramine Hydrochloride, Mikromol™
Artikelnummer. 30458746
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458746 4 x 25 mg
30049739 100 mg
30049908 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458746 Leverandør Mikromol Leverandørnr. MM0608.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Norfenfluramine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Norfenfluramine Hydrochloride
CAS 673-18-7
Analyt- eller komponentnavne Norfenfluramine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C10 H12 F3 N . Cl H
Certificeringer/compliance ISO 9001
Synonym Norfenfluramine Hydrochloride
InChI formel InChI=1S/C10H12F3N.ClH/c1-7(14)5-8-3-2-4-9(6-8)10(11,12)13;/h2-4,6-7H,5,14H2,1H3;1H
Løsningstype Neat
SMIL Cl.CC(N)Cc1cccc(c1)C(F)(F)F
IUPAC navn 1-[3-(trifluoromethyl)phenyl]propan-2-amine;hydrochloride
Molekylvægt (g/mol) 239.67
Mængde 4 x 25 mg
Formel vægt 239.0689 g/mol