Få mere at vide
Norethynodiol 17-Monoacetate, TRC
Artikelnummer. 30415311
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415311 100 mg
30413530 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30415311 Leverandør Toronto Research Chemicals Leverandørnr. TRCN676050100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2061-46-3
Molekylær formel C22H30O3
Molekylvægt (g/mol) 342.47
Synonym (3beta,17alpha)-19-Norpregn-4-en-20-yne-3,17-diol 17-Acetate, 19-Nor-17alpha-pregn-4-en-20-yne-3beta,17-diol 17-Acetate, 17alpha-Ethynyl-3beta-hydroxy-4-estrene-17beta-acetate, SC 12222
IUPAC navn [(3S,8R,9S,10R,13S,14S,17R)-17-ethynyl-3-hydroxy-13-methyl-2,3,6,7,8,9,10,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMIL CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=C[C@@H](O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
Tekniske data
Kemisk navn eller materiale Norethynodiol 17-Monoacetate
Mængde 100 mg
InChI formel InChI=1S/C22H30O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,13,16-20,24H,5-12H2,2-3H3/t16-,17-,18+,19+,20-,21-,22-/m0/s1
Formel vægt 342.2195