missing translation for 'onlineSavingsMsg'
Learn More

Norethindrone, MP Biomedicals™

Catalog Number p-4405164
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
68-22-4
C20H26O2
298.426
VIKNJXKGJWUCNN-XGXHKTLJSA-N
norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
6230
CHEBI:7627
(8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34
This item is not returnable. View return policy
68-22-4
C20H26O2
298.426
VIKNJXKGJWUCNN-XGXHKTLJSA-N
norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
6230
CHEBI:7627
(8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'