Learn More
GPE Scientific™ Norell™ Fluoropolymer Liners
Product Code. 12862316
Change view
Click to view available options
Description:
Capacity: 0.35 mL
Capacity: 2 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
12862316 Capacity: 2 mL Each
12852316 Capacity: 0.35 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12862316 Supplier GPE Scientific™ Supplier No. TL107
  • ONLINE EXCLUSIVE PRICE
    572.00 DKK
    497.55 DKK / Pakke med 1
    Spare 74.45 DKK
    13% Slukket
Forventet forsendelse: 18-12-2026