Få mere at vide
Nordefrin Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Nordefrin Hydrochloride |
| CAS | 61-96-1 |
| Analyt- eller komponentnavne | Nordefrin Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H13 N O3 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1,2-Benzenediol, 4-[(1R,2S)-2-amino-1-hydroxypropyl]-, hydrochloride (1:1), rel-, 1,2-Benzenediol, 4-(2-amino-1-hydroxypropyl)-, hydrochloride, (R*,S*)-(±)-, 1,2-Benzenediol, 4-[(1R,2S)-2-amino-1-hydroxypropyl]-, hydrochloride, rel- (9CI), Benzyl alcohol, α-(1-aminoethyl)-3,4-dihydroxy-, hydrochloride, erythro-(±)- (8CI), (±)-Cobefrin, (±)-Cobefrin hydrochloride, (±)-α-Methylnoradrenaline hydrochloride, (±)-α-Methylnorepinephrine hydrochloride, 1,2-Benzenediol, 4-(2-amino-1-hydroxypropyl)-, hydrochloride, (R*,S*)-, Cobefrin, Corbasil, DL-α-Methylnorepinephrine hydrochloride, Nordefrin hydrochloride, dl-Corbadrine hydrochloride, dl-α-(1-Aminoethyl)-3,4-dihydroxybenzyl alcohol hydrochloride |
| InChI formel | InChI=1S/C9H13NO3.ClH/c1-5(10)9(13)6-2-3-7(11)8(12)4-6;/h2-5,9,11-13H,10H2,1H3;1H/t5-,9-;/m1./s1 |
| Løsningstype | Neat |
| SMIL | Cl.C[C@@H](N)[C@@H](O)c1ccc(O)c(O)c1 |
| Vis mere |