Få mere at vide
Nor Verapamil-d7, Hydrochloride, TRC
Artikelnummer. 30423226
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423226 1 mg
30415056 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423226 Leverandør Toronto Research Chemicals Leverandørnr. TRCN8775031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216413-74-9
Molekylær formel C26 2H7 H29 N2 O4 . Cl H
Molekylvægt (g/mol) 484.08
Synonym α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]amino]propyl]-3,4-dimethoxy-α-[1-(methyl-d3)ethyl-1,2,2,2-d4]-benzeneacetonitrile hydochloride, 2-(3,4-Dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethylamino]-2-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)pentanenitrile hydrochloride, Nor Verapamil-d7 hydrochloride, N-Norverapamil-D7 hydrochloride (isopropyl-D7)
IUPAC navn 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethylamino]-2-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)pentanenitrile;hydrochloride
SMIL Cl.[2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])C(CCCNCCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2
Tekniske data
Kemisk navn eller materiale Norverapamil-d7 hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C26H36N2O4.ClH/c1-19(2)26(18-27,21-9-11-23(30-4)25(17-21)32-6)13-7-14-28-15-12-20-8-10-22(29-3)24(16-20)31-5;/h8-11,16-17,19,28H,7,12-15H2,1-6H3;1H/i1D3,2D3,19D;
Formel vægt 483.2881