Få mere at vide
Nor Propranolol-d7 Hydrochloride, TRC
Artikelnummer. 30422428
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30422428 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422428 Leverandør Toronto Research Chemicals Leverandørnr. TRCN8256022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nor Propranolol-d7 Hydrochloride
CAS 1246819-85-1
Molekylær formel C13 D5 H10 N O2 . H Cl
Mængde 2.5 mg
Synonym 1-Amino-3-(1-naphthalenyloxy)-2-propanol-d7 Hydrochloride, 1-Amino-3-(alpha-naphthyloxy)propan-2-ol-d7 Hydrochloride, (+/-)-Desisopropylpropranolol-d7 Hydrochloride, (+/-)-Norpropranolol-d7 Hydrochloride, N-Deisopropylpropranolol-d7 Hydrochloride, N-Desisopropylpropranolol-d7 Hydrochloride
InChI formel InChI=1S/C13H15NO2.ClH/c14-8-11(15)9-16-13-7-3-5-10-4-1-2-6-12(10)13;/h1-7,11,15H,8-9,14H2;1H/i8D2,9D2,11D;
SMIL Cl.[2H]C([2H])(N)C([2H])(O)C([2H])([2H])Oc1cccc2ccccc12
IUPAC navn 1-amino-1,1,2,3,3-pentadeuterio-3-naphthalen-1-yloxypropan-2-ol;hydrochloride
Molekylvægt (g/mol) 258.755
Formel vægt 258.118