Få mere at vide
Nor NOHA monoacetate, Tocris Bioscience™
Arginase inhibitor
Brand: Tocris Bioscience 6370/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Nor NOHA monoacetate is a reversible, competitive arginase inhibitor (IC50 = 2 μM). Exhibits 10-fold selectivity for human type II arginase over type I. Enhances the effect of acetylcholine on isolated aortic and mesenteric arterial rings. Inhibits growth of lung carcinoma implants in mice.Tekniske data
| (2S)-2-Amino-4-[[(hydroxyamino)iminomethyl]amino]butanoic acid monoacetate | |
| 2250019-93-1 | |
| 10 mg |