Få mere at vide
Nor Neostigmine, TRC
Artikelnummer. 30413848
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30413848 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413848 Leverandør Toronto Research Chemicals Leverandørnr. TRCN824400250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nor Neostigmine
CAS 16088-19-0
Renhedsgrad noter HPLC
Molekylær formel C11H16N2O2
Mængde 250 mg
Synonym Dimethylcarbamic Acid 3-(Dimethylamino)phenyl Ester, Dimethylcarbamic Acid m-(Dimethylamino)phenyl Ester, 3-(Dimethylamino)phenyl Dimethylcarbamate, Norneostigmine
InChI formel InChI=1S/C11H16N2O2/c1-12(2)9-6-5-7-10(8-9)15-11(14)13(3)4/h5-8H,1-4H3
SMIL CN(C)C1=CC(=CC=C1)OC(=O)N(C)C
IUPAC navn [3-(dimethylamino)phenyl] N,N-dimethylcarbamate
Molekylvægt (g/mol) 208.26
Formel vægt 208.1212
Procent renhed >95