Få mere at vide
Nor Mianserin Hydrochloride, TRC
Artikelnummer. 30424799
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424799 5 mg
30422355 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30424799 Leverandør Toronto Research Chemicals Leverandørnr. TRCN7368075MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76134-77-5
Molekylær formel C17 H18 N2 . Cl H
Molekylvægt (g/mol) 286.8
Synonym (14bRS)-1,2,3,4,10,14b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine Hydrochloride, Mianserin Hydrochloride Imp. E (EP) as Hydrochloride, Desmethylmianserin Hydrochloride, Normianserin Hydrochloride
IUPAC navn (14bRS)-1,2,3,4,10,14b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride
SMIL Cl.C1CN2C(CN1)c3ccccc3Cc4ccccc24
Tekniske data
Kemisk navn eller materiale Normianserin Hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C17H18N2.ClH/c1-3-7-15-13(5-1)11-14-6-2-4-8-16(14)19-10-9-18-12-17(15)19;/h1-8,17-18H,9-12H2;1H
Formel vægt 286.1237