missing translation for 'onlineSavingsMsg'
Få mere at vide

Nor Lidocaine Hydrochloride, TRC

Artikelnummer. 30008677
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008677 50 mg
17934845 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30008677 Leverandør Toronto Research Chemicals Leverandørnr. TRCN71350050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 7729-94-4
Molekylær formel C12 H18 N2 O . Cl H
Molekylvægt (g/mol) 242.75
Synonym N-(2,6-Dimethylphenyl)-2-(ethylamino)acetamide Hydrochloride, Lidocaine Hydrochloride Imp. D (EP) as Hydrochloride, Monoethylglycinexylidide Hydrochloride, Norlidocaine Hydrochloride
IUPAC navn N-(2,6-dimethylphenyl)-2-(ethylamino)acetamide;hydrochloride
SMIL Cl.CCNCC(=O)Nc1c(C)cccc1C

Tekniske data

Kemisk navn eller materiale Norlidocaine Hydrochloride (Monoethylglycinexylidide Hydrochloride)
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C12H18N2O.ClH/c1-4-13-8-11(15)14-12-9(2)6-5-7-10(12)3;/h5-7,13H,4,8H2,1-3H3,(H,14,15);1H
Formel vægt 242.1186
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.