Få mere at vide
Nor Harmane-d7, TRC
Artikelnummer. 30410967
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30410967 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410967 Leverandør Toronto Research Chemicals Leverandørnr. TRCN70000210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nor Harmane-d7
CAS 1216503-21-7
Renhedsgrad noter HPLC
Molekylær formel C11HD7N2
Mængde 10 mg
Synonym 9H-Pyrido[3,4-b]indole-d7, β-Carboline-d7, 2,9-Diazafluorene-d7, 2-Azacarbazole-d7, Carbazoline-d7, Norharman-d7, NSC 84417-d7
InChI formel InChI=1S/C11H8N2/c1-2-4-10-8(3-1)9-5-6-12-7-11(9)13-10/h1-7,13H/i1D,2D,3D,4D,5D,6D,7D
SMIL [2H]C1=C([2H])C([2H])=C([2H])C(N2)=C1C3=C2C([2H])=NC([2H])=C3[2H]
IUPAC navn 1,3,4,5,6,7,8-heptadeuterio-9H-pyrido[3,4-b]indole
Molekylvægt (g/mol) 175.24
Formel vægt 175.1127
Procent renhed >95