Få mere at vide
Nor Doxepin-d3 Hydrochloride, TRC
Artikelnummer. 30409473
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30409473 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409473 Leverandør Toronto Research Chemicals Leverandørnr. TRCN67350210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyldoxepin-D3 hydrochloride (cis/trans)
CAS 1331665-54-3
Renhedsgrad noter HPLC
Molekylær formel C18 2H3 H16 N O . Cl H
Mængde 10 mg
Synonym 3-(6H-Benzo[c][2]benzoxepin-11-ylidene)-N-(trideuteriomethyl)propan-1-amine hydrochloride, Nordoxepin-D3 hydrochloride (cis/trans), N-Desmethyldoxepin-D3 hydrochloride (cis/trans), Desmethyldoxepine-D3 (N-methyl-D3) hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-(methyl-d3)propan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine-D3 (N-methyl-D3) hydrochloride
InChI formel InChI=1S/C18H19NO.ClH/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18;/h2-5,7-11,19H,6,12-13H2,1H3;1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])NCCC=C1c2ccccc2COc3ccccc13
IUPAC navn 3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N-(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 304.83
Formel vægt 304.1422
Procent renhed >95