Få mere at vide
Nor Acetildenafil-d8, TRC
Artikelnummer. 30481501
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30481501 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481501 Leverandør Toronto Research Chemicals Leverandørnr. TRCN66050210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nor-acetildenafil-d8
CAS 1185117-07-0
Molekylær formel C24H24D8N6O3
Mængde 10 mg
InChI formel InChI=1S/C24H32N6O3/c1-5-7-18-21-22(29(4)27-18)24(32)26-23(25-21)17-14-16(8-9-20(17)33-6-2)19(31)15-30-12-10-28(3)11-13-30/h8-9,14H,5-7,10-13,15H2,1-4H3,(H,25,26,32)/i10D2,11D2,12D2,13D2
SMIL [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])N(CC(=O)c2ccc(OCC)c(c2)C3=Nc4c(CCC)nn(C)c4C(=O)N3)C1([2H])[2H]
Molekylvægt (g/mol) 460.6
Formel vægt 460.3038