Få mere at vide
Nonoxynol-3 Phosphate, TRC
Artikelnummer. 30471799
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471799 1 g
30468148 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30471799 Leverandør Toronto Research Chemicals Leverandørnr. TRCN6495731G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nonoxynol-3 Phosphate
CAS 51811-79-1
Molekylær formel C18 H30 O . x (H3 O4 P)
Mængde 1 g
Synonym Cedephos CP 610, Gafac, Monafax, Plysurf, Soprophor, Polyoxyethylene nonylphenyl ether phosphate
InChI formel InChI=1S/C18H30O.H3O4P/c1-2-3-4-5-6-7-8-12-17-13-9-10-14-18(17)15-11-16-19;1-5(2,3)4/h9-10,13-14,19H,2-8,11-12,15-16H2,1H3;(H3,1,2,3,4)
SMIL CCCCCCCCCc1ccccc1CCCO.OP(=O)(O)O
IUPAC navn 3-(2-nonylphenyl)propan-1-ol;phosphoric acid