Få mere at vide
Nonanedioic-D14 Acid, TRC
Artikelnummer. 30467798
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30467798 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467798 Leverandør Toronto Research Chemicals Leverandørnr. TRCN64934710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nonanedioic-D14 Acid
CAS 119176-67-9
Molekylær formel C9 2H14 H2 O4
Mængde 10 mg
Synonym Nonanedioic-d14 acid (9CI), 1,9-Nonanedioic-d14 Acid, Nonanedioic Acid-D14, Azelaic acid-d14
InChI formel InChI=1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2
SMIL [2H]C([2H])(C(=O)O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecadeuteriononanedioic acid
Molekylvægt (g/mol) 202.31
Formel vægt 202.1927