Få mere at vide
Nonanal-d18, TRC
Artikelnummer. 30470803
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30470803 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470803 Leverandør Toronto Research Chemicals Leverandørnr. TRCN64933210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nonanal-d18
CAS 1466552-36-2
Molekylær formel C9 D18 O
Mængde 10 mg
Synonym Nonanaldehyde-d18, Aldehyde C 09-d18, Aldehyde C 9-d18, NSC 5518-d18, Nonaldehyde-d18, Nonanoic Aldehyde-d18, Nonylaldehyde-d18, Nonylic Aldehyde-d18, Pelargonaldehyde-d18, Pelargonic Aldehyde-d18, n-Nonanal-d18, n-Nonylaldehyde-d18
InChI formel InChI=1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D
SMIL [2H]C(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-octadecadeuteriononan-1-one
Molekylvægt (g/mol) 160.35
Formel vægt 160.2487