Få mere at vide
Greiner Bio-One Non-binding Surface 384-well Microplates
Beskrivelse
Characteristic features of the non-binding surface are:
Advantages: significantly increase assay sensitivity--reducing background--improving signal-to-noise ratio
- Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
Advantages: significantly increase assay sensitivity--reducing background--improving signal-to-noise ratio
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Can be traced all the way back to production through a defined LOT number system
- Well profile: Small Volume™
Tekniske data
Tekniske data
| Stregkode | Alphanumeric |
| Certificeringer/compliance | DIN ISO 9001 |
| Format | Standard |
| Array | 16 x 24 |
| Indbindingstype | Non Binding |
| Farve | Sort |
| Materiale | Polystyrene |
| Antal Wells | 384 |
| Sterilitet | Non-sterile |
| Overfladebehandling | Protein-Repellent Non-binding |
| Vis mere |