missing translation for 'onlineSavingsMsg'
Få mere at vide

Nomifensine Maleic Acid Salt, TRC

Product Code. 30483164
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30483164 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483164 Supplier Toronto Research Chemicals Supplier No. TRCN635950100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Nomifensine Hydrogen Maleate
CAS 32795-47-4
Molekylær formel C16 H18 N2 . C4 H4 O4
Mængde 100 mg
Synonym 1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine (2Z)-2-butenedioate (1:1), 1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine (Z)-2-butenedioate (1:1), (±)-Nomifensine hydrogen maleate, Alival, Alival (antidepressant), HOE 984, Hostalival, Merital, NSC 289114, Neurolene, Nomifensine hydrogen maleate, Nomifensine maleate, Psicronizer
InChI formel InChI=1S/C16H18N2.C4H4O4/c1-18-10-14(12-6-3-2-4-7-12)13-8-5-9-16(17)15(13)11-18;5-3(6)1-2-4(7)8/h2-9,14H,10-11,17H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMIL CN1CC(c2ccccc2)c3cccc(N)c3C1.OC(=O)\C=C/C(=O)O
IUPAC navn (Z)-but-2-enedioic acid;2-methyl-4-phenyl-3,4-dihydro-1H-isoquinolin-8-amine
Molekylvægt (g/mol) 354.4
Formel vægt 354.158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.