Få mere at vide
Nodakenin, TRC
Artikelnummer. 30478382
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30478382 5 mg -
30477315 2.5 mg 2.5mg
30483487 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30478382 Leverandør Toronto Research Chemicals Leverandørnr. TRCN2318355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 495-31-8
Molekylær formel C20 H24 O9
Molekylvægt (g/mol) 408.4
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one, 2-[1-(β-D-glucopyranosyloxy)-1-methylethyl]-2,3-dihydro-, (2R)-, 7H-Furo[3,2-g][1]benzopyran-7-one, 2-[1-(β-D-glucopyranosyloxy)-1-methylethyl]-2,3-dihydro-, (R)-, Nodakenin (8CI), (+)-Marmesinin, (2R)-2-[1-(β-D-Glucopyranosyloxy)-1-methylethyl]-2,3-dihydro-7H-furo[3,2-g][1]benzopyran-7-one, Nodakenetin, β-D-glucopyranoside
IUPAC navn (2R)-2-[2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxypropan-2-yl]-2,3-dihydrofuro[3,2-g]chromen-7-one
SMIL CC(C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2Cc3cc4C=CC(=O)Oc4cc3O2
Tekniske data
Kemisk navn eller materiale Nodakenin
Mængde 5 mg
InChI formel InChI=1S/C20H24O9/c1-20(2,29-19-18(25)17(24)16(23)13(8-21)28-19)14-6-10-5-9-3-4-15(22)27-11(9)7-12(10)26-14/h3-5,7,13-14,16-19,21,23-25H,6,8H2,1-2H3/t13-,14-,16-,17+,18-,19+/m1/s1
Formel vægt 408.142