Få mere at vide
Nociceptin Trifluoroacetic Acid Salt, TRC
Artikelnummer. 30136479
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136479 5 mg
30135717 1 mg
30010252 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136479 Leverandør Toronto Research Chemicals Leverandørnr. TRCN6341005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 218297-67-7
Molekylær formel C79H129N27O22 . x(C 2HF3O2)
Molekylvægt (g/mol) 1923.1
Synonym L-Phenylalanylglycylglycyl-L-phenylalanyl-L-threonylglycyl-L-alanyl-L-arginyl-L-lysyl-L-seryl-L-alanyl-L-arginyl-L-lysyl-L-leucyl-L-alanyl-L-asparaginyl-L-glutamine, Orphanin FQ (pig), Orphanin FQ (swine), 102: PN: US20090104234 SEQID: 102 unclaimed protein, 10: PN: WO2007138336 SEQID: 2 claimed sequence, 114: PN: US20030119021 SEQID: 103 unclaimed sequence TFA Salt, 12: PN: WO2008011381 SEQID: 68 unclaimed sequence, 133: PN: US20120207734 SEQID: 132 claimed sequence, 13: PN: WO2004073646 TABLE: 3 unclaimed sequence, 142: PN: WO2008125902 SEQID: 132 unclaimed sequence, 154: PN: US20090048431 SEQID: 153 claimed sequence, 15: PN: WO03093294 SEQID: 1 claimed protein, 174: PN: US20110189162 SEQID: 174 unclaimed sequence, 1: PN: US20110021426 SEQID: 1 claimed protein, 1: PN: US20120207735 SEQID: 2 claimed protein, 1: PN: WO2006059105 SEQID: 2 claimed sequence, 241: PN: WO0069900 SEQID: 919 unclaimed protein, 246: PN: WO0069900 SEQID: 926 unclaimed protein, 24: PN: US20030045696 SEQID: 24 unclaimed sequence, 24: PN: US20050169917 SEQID: 24 unclaimed sequence, 2: PN: US20120058098 SEQID: 2 unclaimed protein, 2: PN: US20120156186 SEQID: 2 unclaimed sequence, 2: PN: US20120230975 SEQID: 38 claimed protein, 38: PN: US20110027256 SEQID: 38 unclaimed sequence, 42: PN: WO2009012472 SEQID: 42 unclaimed sequence, 44: PN: WO2006068768 SEQID: 42 claimed sequence, 46: PN: WO2007138339 SEQID: 38 claimed sequence, 51: PN: WO2011020052 SEQID: 132 claimed protein, 52: PN: WO2008008805 SEQID: 53 unclaimed protein, 730: PN: US20090175821 SEQID: 919 claimed protein, 79: PN: WO2011146704 SEQID: 76 unclaimed protein, 86: PN: WO03097857 PAGE: 39 claimed protein, 87: PN: WO03097857 PAGE: 39 claimed protein, 90: PN: US20110091437 SEQID: 38 unclaimed sequence TFA Salt, 9: PN: WO2008052043 SEQID: 8 claimed protein TFA Salt, Human nociceptin TFA Salt, Nociceptin (human) TFA Salt, Nociceptin 1-17, Orphanin FQ (human), Orphanin FQ (rat) TFA Salt
SMIL O=C(N)CC[C@@H](C(O)=O)NC([C@H](CC(N)=O)NC([C@H](C)NC([C@H](CC(C)C)NC([C@H](CCCCN)NC([C@H](CCCNC(N)=N)NC([C@H](C)NC([C@H](CO)NC([C@H](CCCCN)NC([C@H](CCCNC(N)=N)NC([C@H](C)NC(CNC([C@H]([C@H](O)C)NC([C@H](CC1=CC=CC=C1)NC(CNC(CNC([C@H](CC2=CC=CC=C2)N)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O.FC(F)(C(O)=O)F
Tekniske data
Kemisk navn eller materiale Nociceptin Trifluoroacetic Acid Salt
Mængde 5 mg
InChI formel InChI=1S/C79H129N27O22.C2HF3O2/c1-41(2)33-54(72(122)95-44(5)66(116)103-56(36-59(84)110)73(123)102-53(77(127)128)27-28-58(83)109)104-70(120)49(23-13-15-29-80)100-69(119)52(26-18-32-90-79(87)88)99-65(115)43(4)96-75(125)57(40-107)105-71(121)50(24-14-16-30-81)101-68(118)51(25-17-31-89-78(85)86)98-64(114)42(3)94-61(112)39-93-76(126)63(45(6)108)106-74(124)55(35-47-21-11-8-12-22-47)97-62(113)38-91-60(111)37-92-67(117)48(82)34-46-19-9-7-10-20-46;3-2(4,5)1(6)7/h7-12,19-22,41-45,48-57,63,107-108H,13-18,23-40,80-82H2,1-6H3,(H2,83,109)(H2,84,110)(H,91,111)(H,92,117)(H,93,126)(H,94,112)(H,95,122)(H,96,125)(H,97,113)(H,98,114)(H,99,115)(H,100,119)(H,101,118)(H,102,123)(H,103,116)(H,104,120)(H,105,121)(H,106,124)(H,127,128)(H4,85,86,89)(H4,87,88,90);(H,6,7)/t42?,43?,44?,45?,48-,49-,50-,51-,52-,53-,54-,55-,56-,57-,63-;/m0./s1