Learn More
Schmidlin™ NM Plus Hydrogen Generator
This generator produces a continuous supply of high purity, dry hydrogen.
Brand:  Schmidlin™ DB-PNM1000-EU
Additional Details : Weight : 17.00000kg
 View more versions of this product
Product Code. 17718919
  • 186534.58 DKK / Each
Estimated Shipment: 31-07-2024
to see stock.
Description
Description
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Pressure: 11 barg (160 psig)
  • Unique permeation membrane drying system
  • Cold dual dynamic regeneration dryer
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Meets and exceeds requirements for the most demanding GC applications
  • Superior hydrogen production with reliable long life cell
  • Minimal maintenance; no desiccant cartridges to change
  • PC monitoring for maintenance, diagnostics and remote connection
Specifications
Specifications
37 cm
50.7 cm
11 bar
23 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
385 W
1000 mL/min.
35°C
9 in.
Outlet: 1/8 in.
>99.99996%