Få mere at vide
Nizatidine, TRC
Artikelnummer. 30480071
Skift visning
Mængde:
50 mg
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480071 50 mg
30465489 1 g
30478997 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480071 Leverandør Toronto Research Chemicals Leverandørnr. TRCN59850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76963-41-2
Molekylær formel C12 H21 N5 O2 S2
Molekylvægt (g/mol) 331.46
Synonym Nizatidine, N'-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N-methyl-2-nitro-1,1-ethenediamine, N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine, Acinon, Axid, Bonacid, Calmaxid, Cronizat, Distaxid, Gastrax, LY 139037, Naxidine, Nizax, Nizaxid, Tazac, ZL 101, Zanizal
IUPAC navn 1-N'-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
SMIL CNC(=C[N+](=O)[O-])NCCSCc1csc(CN(C)C)n1
Tekniske data
Kemisk navn eller materiale Nizatidine
Mængde 50 mg
InChI formel InChI=1S/C12H21N5O2S2/c1-13-11(6-17(18)19)14-4-5-20-8-10-9-21-12(15-10)7-16(2)3/h6,9,13-14H,4-5,7-8H2,1-3H3
Formel vægt 331.1137