Få mere at vide
Nizatidine EP Impurity A, Mikromol™
Artikelnummer. 30051087
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051087 100 mg
30051857 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051087 Leverandør Mikromol Leverandørnr. MM0220.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nizatidine EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N'-Dimethyl-2-nitroethene-1,1-diamine
CAS 54252-45-8
Analyt- eller komponentnavne N,N'-Dimethyl-2-nitroethene-1,1-diamine
Forsendelsestilstand Room Temperature
Molekylær formel C4 H9 N3 O2
Certificeringer/compliance ISO 9001
Synonym N,N'-Dimethyl-2-nitroethene-1,1-diamine, Nizatidine Imp. A (EP), Nizatidine Impurity A
InChI formel InChI=1S/C4H9N3O2/c1-5-4(6-2)3-7(8)9/h3,5-6H,1-2H3
SMIL CNC(=C[N+](=O)[O-])NC
IUPAC navn 1-N,1-N'-dimethyl-2-nitroethene-1,1-diamine
Molekylvægt (g/mol) 131.13
Mængde 100 mg
Formel vægt 131.0695 g/mol