Få mere at vide
Nizatidine-d3, TRC
Artikelnummer. 30482359
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30482359 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482359 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5985021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nizatidine-d3
CAS 1246833-99-7
Molekylær formel C12 D3 H18 N5 O2 S2
Mængde 1 mg
Synonym N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N’-(methyl-d3)-2-nitro-1,1-ethenediamine, LY-139037-d3, ZE-101-d3, ZL-101-d3, Axid-d3, Calmaxid-d3, Cronizat-d3, Distaxid-d3, Gastrax-d3, Nizax-d3, Nizaxid-d3
InChI formel InChI=1S/C12H21N5O2S2/c1-13-11(6-17(18)19)14-4-5-20-8-10-9-21-12(15-10)7-16(2)3/h6,9,13-14H,4-5,7-8H2,1-3H3/b11-6+/i1D3
SMIL [2H]C([2H])([2H])N\C(=C/[N+](=O)[O-])\NCCSCc1csc(CN(C)C)n1
IUPAC navn (E)-N1-[2-[[2-[(dimethylamino)methyl]thiazol-4-yl]methylsulfanyl]ethyl]-2-nitro-N1'-(trideuteriomethyl)ethene-1,1-diamine
Molekylvægt (g/mol) 334.48
Formel vægt 334.1325