Få mere at vide
Nitrofurantoin Monohydrate, TRC
Artikelnummer. 30476684
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476684 100 mg
30472118 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30476684 Leverandør Toronto Research Chemicals Leverandørnr. TRCN075910100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17140-81-7
Molekylær formel C8H8N4O6
Molekylvægt (g/mol) 256.18
Synonym 1-(((5-Nitrofuran-2-yl)methylene)amino)-imidazolidine-2,4-dione Hydrate, Furadantin Monohydrate
IUPAC navn 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]imidazolidine-2,4-dione;hydrate
SMIL O.[O-][N+](=O)c1oc(\C=N\N2CC(=O)NC2=O)cc1
Tekniske data
Kemisk navn eller materiale Nitrofurantoin Monohydrate
Mængde 100 mg
InChI formel InChI=1S/C8H6N4O5.H2O/c13-6-4-11(8(14)10-6)9-3-5-1-2-7(17-5)12(15)16;/h1-3H,4H2,(H,10,13,14);1H2/b9-3+;
Formel vægt 256.0444