missing translation for 'onlineSavingsMsg'
Learn More

Nitroethane, ≥98.0%, Reagent Grade, Honeywell™

Brand:  Honeywell Research Chemicals 130206-1L

Additional Details : CAS Number : 79-24-3 Weight : 1.04500kg Transport : UN number : 2842 Chem class : 3 Pack group : III

Packaging Glass bottle
Quantity 1L

 View more versions of this product

Product Code. 15641240

  • 567.38 DKK / 1 L


Estimated Shipment date
from Supplier 09-08-2018
Add to basket

Specifications

Chemical Identifiers

Slightly soluble in water, Soluble in acetone, Soluble in alcohol
15.6mmHg (20°C)
31°C (87.8°F)
CH3CH2NO2
UN2842
75.07g/mol
Nitroethane
1209324
−90°c
2.58 (vs air)
1.045g/mL at 25°C
778°F
MFCD00007404
≥98.0%
Reagent grade
79-24-3
114° to 116°C
1.391

Specifications

Nitroethane
Reagent grade
CH3CH2NO2
MFCD00007404
1209324
778°F
1.391
15.6mmHg (20°C)
114° to 116°C
1L
79-24-3
≥98.0%
75.07g/mol
UN2842
1.045g/mL at 25°C
Glass bottle
Slightly soluble in water, Soluble in acetone, Soluble in alcohol
2.58 (vs air)
−90°c
31°C (87.8°F)
Videos